Learn More
Medchemexpress LLC Methapyrilene hydrochloride | 135-23-9 | MFCD00050665 | 99.9% | 297.85 g/mol | C14H20ClN3S | 10 MG
SDP
Catalog No. 5000297986
Encompass_Preferred
Methapyrilene hydrochloride is a research-grade H1-receptor antihistamine with anticholinergic activity, supplied as a solid for pharmacology and toxicology studies. It has documented hepatotoxic effects and is used experimentally to model periportal hepatic necrosis. Chemical formula C14H20ClN3S, molecular weight 297.85 g/mol, purity 99.9%.
  • H1-receptor antihistamine with anticholinergic activity.
  • Used in pharmacology and toxicology research and liver injury models.
  • High purity solid suitable for analytical and in vivo studies.
  • CAS number 135-23-9 for unambiguous identification.
  • Store sealed, away from moisture; in solution store at -20°C short term or -80°C long term.
Catalog No. 50-002-97986 Supplier Medchemexpress LLC Supplier No. HYB148310MG
Methapyrilene hydrochloride is a research-grade H1-receptor antihistamine with anticholinergic activity, supplied as a solid for pharmacology and toxicology studies. It has documented hepatotoxic effects and is used experimentally to model periportal hepatic necrosis. Chemical formula C14H20ClN3S, molecular weight 297.85 g/mol, purity 99.9%.
  • H1-receptor antihistamine with anticholinergic activity.
  • Used in pharmacology and toxicology research and liver injury models.
  • High purity solid suitable for analytical and in vivo studies.
  • CAS number 135-23-9 for unambiguous identification.
  • Store sealed, away from moisture; in solution store at -20°C short term or -80°C long term.