Learn More
Cayman Chemical MethapyrIlenhydrochLRIde 100mg
SDP
Catalog No. 502947402
Encompass_Preferred
An H1 receptor antagonist (Ki = 4.5 nM) and non-genotoxic carcinogen; induces liver tumors in rats at 0.1% in the diet; decreases H3K9 and H3K56 acetylation, as well as H3K4 di- and H4K20 trimethylation, in rat liver at 40 mg/kg per day for six weeks
Catalog No. 50-294-7402 Supplier Cayman Chemical Supplier No. 33434100 MG
An H1 receptor antagonist (Ki = 4.5 nM) and non-genotoxic carcinogen; induces liver tumors in rats at 0.1% in the diet; decreases H3K9 and H3K56 acetylation, as well as H3K4 di- and H4K20 trimethylation, in rat liver at 40 mg/kg per day for six weeks