Learn More
Apexbio Technology LLC Methazolamide 554-57-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062257
Encompass_Preferred
Methazolamide (CAS 554-57-4) is a sulfonamide-derived inhibitor of carbonic anhydrase (CA) selectively targeting isoforms CAII and CAIV with IC50 values of 8 1 nM and 80 3 nM respectively By suppressing CA activity methazolamide reduces bicarbonate ion production thus decreasing fluid secretion and intraocular pressure consequently it has clinical use in glaucoma management Recent studies indicate this compound also exhibits insulin-sensitizing properties distinct from other CA inhibitors notably reducing hepatic glucose output suggesting potential applications in diabetes research and metabolic disorders
Catalog No. 50-000-62257 Supplier Apexbio Technology LLC Supplier No. A43645.1
Methazolamide (CAS 554-57-4) is a sulfonamide-derived inhibitor of carbonic anhydrase (CA) selectively targeting isoforms CAII and CAIV with IC50 values of 8 1 nM and 80 3 nM respectively By suppressing CA activity methazolamide reduces bicarbonate ion production thus decreasing fluid secretion and intraocular pressure consequently it has clinical use in glaucoma management Recent studies indicate this compound also exhibits insulin-sensitizing properties distinct from other CA inhibitors notably reducing hepatic glucose output suggesting potential applications in diabetes research and metabolic disorders