Learn More
Selleck Chemical LLC Methazolamide 50mg 554-57-4 CL 8490
SDP
Catalog No. 501363580
Encompass_Preferred
Methazolamide (CL 8490) is a carbonic anhydrase inhibitor with Ki of 50 nM, 14 nM and 36 nM for hCA I, hCA II and bCA IV isoforms, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3580 Supplier Selleck Chemical LLC Supplier No. S403950MG
Methazolamide (CL 8490) is a carbonic anhydrase inhibitor with Ki of 50 nM, 14 nM and 36 nM for hCA I, hCA II and bCA IV isoforms, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.