Learn More
Apexbio Technology LLC Methazolamide, 50mg. Cas: 554-57-4 MFCD: MFCD00083416. Carbonic anhydrase inhibitor
SDP
Catalog No. 501904930
Encompass_Preferred
Methazolamide, a derivative of sulfonamide, is an inhibitor of carbonic anhydrase (CA) that potently inhibits isoenzymes of CA, CA II and CA IV with values of 50% inhibition concentration IC50 of 8.1 nM and 80.3 nM respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4930 Supplier Apexbio Technology LLC Supplier No. A436450
Methazolamide, a derivative of sulfonamide, is an inhibitor of carbonic anhydrase (CA) that potently inhibits isoenzymes of CA, CA II and CA IV with values of 50% inhibition concentration IC50 of 8.1 nM and 80.3 nM respectively. Other sizes are available. Please inqury us for quote.