Learn More
Cayman Chemical Methedrone (hydrochloride), Purity: ≥98%, CAS: 879665-92-6, 50 mg
SDP
Catalog No. 502890501
Encompass_Preferred
A phenethylamine, amphetamine, and cathinone derivative that acts as a triple reuptake/release/reversible MAO inhibitor; intended for use as a standard for the forensic analysis of samples that may contain this compound
Catalog No. 50-289-0501 Supplier Cayman Chemical Supplier No. 1052950 MG
A phenethylamine, amphetamine, and cathinone derivative that acts as a triple reuptake/release/reversible MAO inhibitor; intended for use as a standard for the forensic analysis of samples that may contain this compound