Learn More
Cayman Chemical Methenamine Hippurate, Purity: ≥98%, CAS: 5714-73-8, 100 mg
SDP
Catalog No. 502933033
Encompass_Preferred
A broad-spectrum urinary antiseptic agent; is hydrolyzed to formaldehyde and ammonia, which exerts an antibacterial effect under acidic conditions; is effective against a wide range of bacteria but lacks activity against Proteus and other bacteria that increase urine alkalinity
Catalog No. 50-293-3033 Supplier Cayman Chemical Supplier No. 24004100 MG
A broad-spectrum urinary antiseptic agent; is hydrolyzed to formaldehyde and ammonia, which exerts an antibacterial effect under acidic conditions; is effective against a wide range of bacteria but lacks activity against Proteus and other bacteria that increase urine alkalinity