Learn More
Electron Microscopy Sciences Methenamine, 3% Aqueous Solution 500 ML
SDP
Catalog No. 5027159
Encompass_Preferred
A stock solution for use in bacterial and fungal stains (Grocott's Method/GMS). It is a ready-to-use, high quality solution that saves time and effort and provides consistently uniform results.
Catalog No. 50-271-59 Supplier Electron Microscopy Sciences Supplier No. 2608305
A stock solution for use in bacterial and fungal stains (Grocott's Method/GMS). It is a ready-to-use, high quality solution that saves time and effort and provides consistently uniform results.