Learn More
Electron Microscopy Sciences Methenamine, 3% Aqueous Solution 1 LT
SDP
Catalog No. 5027160
Encompass_Preferred
A stock solution for use in bacterial and fungal stains (Grocott's Method/GMS). It is a ready-to-use, high quality solution that saves time and effort and provides consistently uniform results.
Catalog No. 50-271-60 Supplier Electron Microscopy Sciences Supplier No. 2608306
A stock solution for use in bacterial and fungal stains (Grocott's Method/GMS). It is a ready-to-use, high quality solution that saves time and effort and provides consistently uniform results.