Learn More
Electron Microscopy Sciences Methenamine 30% 500 ML
SDP
Catalog No. 5031871
Encompass_Preferred
Used in Gomori's Methenamine Silver for Urate Crystals Urate crystals. Sheehan, Dezna C. and Hrapchak, Barbara B.: Theory and Practice of Histotechnology, 2nd Ed. The C.V. Mosby Company, St. Louis, p.225-226, c. 1980.
Catalog No. 50-318-71 Supplier Electron Microscopy Sciences Supplier No. 2661402
Used in Gomori's Methenamine Silver for Urate Crystals Urate crystals. Sheehan, Dezna C. and Hrapchak, Barbara B.: Theory and Practice of Histotechnology, 2nd Ed. The C.V. Mosby Company, St. Louis, p.225-226, c. 1980.