Learn More
Selleck Chemical LLC Methenamine Hippurate S9466-1MG
SDP
Catalog No. 5000024109
Encompass_Preferred
Methenamine Hippurate is the hippurate salt form of methenamine a prodrug and inactive weak base that is hydrolyzed to formaldehyde in acid urine Methenamine Hippurate is the component of Hiprex drug which has antibacterial activity
Catalog No. 50-000-24109 Supplier Selleck Chemical LLC Supplier No. S94661MG
Methenamine Hippurate is the hippurate salt form of methenamine a prodrug and inactive weak base that is hydrolyzed to formaldehyde in acid urine Methenamine Hippurate is the component of Hiprex drug which has antibacterial activity