Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methenamine hippurate | 5714-73-8 | MFCD00072147 | >95.0% | 319.36 g/mol | C15H21N5O3 | 10 MG
SDP

Catalog No. 5000298900
Encompass_Preferred

Methenamine hippurate is the analytical reference standard of methenamine hippurate supplied for research and analytical use. The compound is the hippurate salt of methenamine, an orally active urinary antiseptic that releases formaldehyde under acidic conditions; supplied as a dry solid and handled per SDS storage recommendations.

  • Analytical reference standard suitable for research and quality control.
  • CAS number 5714-73-8; molecular weight 319.36 g/mol.
  • Purity reported by suppliers as ≥95% (HPLC) to ≥98%; check lot-specific certificate.
  • Recommended storage: 4°C sealed and dry; in solution -80°C (6 months) or -20°C (1 month).
  • Provided as a solid for analytical preparation and formulation testing.

Catalog No. 50-002-98900 Supplier Medchemexpress LLC Supplier No. HYB1691R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methenamine hippurate is the analytical reference standard of methenamine hippurate supplied for research and analytical use. The compound is the hippurate salt of methenamine, an orally active urinary antiseptic that releases formaldehyde under acidic conditions; supplied as a dry solid and handled per SDS storage recommendations.

  • Analytical reference standard suitable for research and quality control.
  • CAS number 5714-73-8; molecular weight 319.36 g/mol.
  • Purity reported by suppliers as ≥95% (HPLC) to ≥98%; check lot-specific certificate.
  • Recommended storage: 4°C sealed and dry; in solution -80°C (6 months) or -20°C (1 month).
  • Provided as a solid for analytical preparation and formulation testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.