Learn More
Medchemexpress LLC Methenamine hippurate (hexamine hippurate) | 5714-73-8 | 99.6% | 100 MG
SDP
Catalog No. 5000413150
Encompass_Preferred
Methenamine hippurate (Hexamine hippurate) is an orally active urinary antiseptic agent with a wide antibacterial spectrum. It is effective against most common urinary tract pathogens. This urinary antibacterial agent releases formaldehyde and is excreted in the urine, demonstrating activity in vitro at pH 5 to 6 against various bacteria.
  • Orally active urinary antiseptic agent.
  • Wide antibacterial spectrum.
  • Effective against most common urinary tract pathogens.
  • Releases formaldehyde in urine.
  • Demonstrates in vitro activity against multiple bacteria including Escherichia coli and Staphylococcus aureus.
Catalog No. 50-004-13150 Supplier Medchemexpress LLC Supplier No. HYB1691100MG
Methenamine hippurate (Hexamine hippurate) is an orally active urinary antiseptic agent with a wide antibacterial spectrum. It is effective against most common urinary tract pathogens. This urinary antibacterial agent releases formaldehyde and is excreted in the urine, demonstrating activity in vitro at pH 5 to 6 against various bacteria.
  • Orally active urinary antiseptic agent.
  • Wide antibacterial spectrum.
  • Effective against most common urinary tract pathogens.
  • Releases formaldehyde in urine.
  • Demonstrates in vitro activity against multiple bacteria including Escherichia coli and Staphylococcus aureus.