Learn More
StatLab Methenamine-Silver Compound
SDP
Catalog No. 22000049
Change view
Click to view available options
10 Vials
Catalog No.
22-000-049 10 Vials
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-049 Supplier StatLab Supplier No. KT093C
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Harmful if swallowed, in contact with skin, or if inhaled
Stains in just fifteen minutes
  • MasterTech Methenamine-Silver Compound Stain Kit is compatible with all G.M.S. Stain Kits
  • Each vial makes 50ml working solution when waters is added
  • Sodium Borate Solution is provided
  • A MasterTech Special Stain Kit
Specifications
Product Type Methenamine/Borax