Learn More
Medchemexpress LLC Methicillin sodium salt | 132-92-3 | 98.1% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB097410MM/1ML
Encompass_Preferred
Methicillin sodium salt is a β-lactam, semi-synthetic antibiotic related to penicillin antibiotics. It inhibits penicillin-binding proteins involved in the synthesis of peptidoglycan. This compound inhibits *S. aureus* with a MIC value of 2.1 μg/mL and is suitable for inflammation research.
  • Semi-synthetic β-lactam antibiotic
  • Inhibits penicillin-binding proteins
  • Effectively eliminates S. aureus
  • Suitable for inflammation research
  • Improves survival in infected animal models
Catalog No. 50-004-08126