Learn More
Research Products International Corp Methicillin Sodium Salt, 1 Gram
SDP
Catalog No. 50488814
Encompass_Preferred
Methicillin Sodium Salt is a chemical compound that belongs to the class of beta-lactam antibiotics known as penicillin.
It is a modified form of penicillin that was developed to be resistant to the effects of the enzyme penicillinase, which is produced by certain bacteria. This resistance allows methicillin to be effective against bacteria that have developed resistance to traditional penicillin antibiotics.
This chemical is primarily used as a selective agent in laboratory experiments to isolate and study bacteria that are resistant to methicillin and other related antibiotics. It is commonly used in microbiology research, particularly in studies focused on antibiotic resistance mechanisms and bacterial genetics.
Researchers may utilize Methicillin Sodium Salt in various techniques such as bacterial culture, antibiotic susceptibility testing, and identification of antibiotic resistance genes.
Catalog No. 50-488-814 Supplier Research Products International Corp Supplier No. M628001.0
Methicillin Sodium Salt is a chemical compound that belongs to the class of beta-lactam antibiotics known as penicillin.
It is a modified form of penicillin that was developed to be resistant to the effects of the enzyme penicillinase, which is produced by certain bacteria. This resistance allows methicillin to be effective against bacteria that have developed resistance to traditional penicillin antibiotics.
This chemical is primarily used as a selective agent in laboratory experiments to isolate and study bacteria that are resistant to methicillin and other related antibiotics. It is commonly used in microbiology research, particularly in studies focused on antibiotic resistance mechanisms and bacterial genetics.
Researchers may utilize Methicillin Sodium Salt in various techniques such as bacterial culture, antibiotic susceptibility testing, and identification of antibiotic resistance genes.