Learn More
Alkali Scientific Methicillin Sodium Salt, 5 G
SDP
Catalog No. 5000659781
Encompass_Preferred
Methicillin Sodium Salt is a beta-lactam antibiotic used primarily to treat infections caused by staphylococci. The 5g powder form is designed for use in laboratory settings, where it is used to prepare solutions for research applications. Methicillin acts by inhibiting bacterial cell wall synthesis, which leads to cell death. It is particularly effective against methicillin-sensitive Staphylococcus aureus (MSSA). This compound is a critical tool for research in antibiotic resistance and bacterial infections. The sodium salt form allows for better solubility in aqueous solutions, making it ideal for in vitro studies and microbiological experiments requiring a specific antibiotic activity.
Catalog No. 50-006-59781 Supplier Alkali Scientific Supplier No. ETH002CP
Methicillin Sodium Salt is a beta-lactam antibiotic used primarily to treat infections caused by staphylococci. The 5g powder form is designed for use in laboratory settings, where it is used to prepare solutions for research applications. Methicillin acts by inhibiting bacterial cell wall synthesis, which leads to cell death. It is particularly effective against methicillin-sensitive Staphylococcus aureus (MSSA). This compound is a critical tool for research in antibiotic resistance and bacterial infections. The sodium salt form allows for better solubility in aqueous solutions, making it ideal for in vitro studies and microbiological experiments requiring a specific antibiotic activity.