Learn More
Medchemexpress LLC HY-B0974 50mg Medchemexpress, Methicillin (sodium salt) CAS:132-92-3 Purity:>98%
SDP
Catalog No. 501970131
Encompass_Preferred
Medchemexpress, HY-B0974 50mg Methicillin (sodium salt) CAS:132-92-3 Methicillin sodium salt (Meticillin sodium) is a β-lactam antibiotic which acts by inhibiting penicillin-binding proteins that are involved in the synthesis of peptidoglycan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0131 Supplier Medchemexpress LLC Supplier No. HYB097450MG
Medchemexpress, HY-B0974 50mg Methicillin (sodium salt) CAS:132-92-3 Methicillin sodium salt (Meticillin sodium) is a β-lactam antibiotic which acts by inhibiting penicillin-binding proteins that are involved in the synthesis of peptidoglycan. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.