Learn More
Cayman Chemical Methimazole, Purity: ≥98%, CAS: 60-56-0, 100 mg
SDP
Catalog No. 502994753
Encompass_Preferred
An inhibitor of thyroid hormone synthesis; a substrate of thyroid peroxidase; traps oxidized iodide and prevents its use by thyroglobulin; inhibits absorption of radiolabeled iodide by the thyroid gland in rats at 0.4 mg/kg; reduces the incidence of lymphocytic thyroiditis in the insulin-dependent type 1 diabetic BB/W rat; has been used to induce hypothyroidism in mice
Catalog No. 50-299-4753 Supplier Cayman Chemical Supplier No. 23718100 MG
An inhibitor of thyroid hormone synthesis; a substrate of thyroid peroxidase; traps oxidized iodide and prevents its use by thyroglobulin; inhibits absorption of radiolabeled iodide by the thyroid gland in rats at 0.4 mg/kg; reduces the incidence of lymphocytic thyroiditis in the insulin-dependent type 1 diabetic BB/W rat; has been used to induce hypothyroidism in mice