Learn More
Medchemexpress LLC HY-B0208 500mg Medchemexpress, Methimazole CAS:60-56-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0208500MG
Encompass_Preferred
Medchemexpress, HY-B0208 500mg Methimazole CAS:60-56-0 Methimazole is an antithyroid compound widely used for the research of hyperthyroidism. Methimazole has potent hepatotoxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2408