Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methimazole | 60-56-0 | 100.0% | 1 ML
SDP

Catalog No. 5000405773
Encompass_Preferred

Methimazole, also known as Thiamazole, is an anti-thyroid agent that effectively blocks the production of thyroid hormones from the thyroid gland. This compound is known to potentially cause hepatotoxicity. It is provided as a 10mM solution in 1mL of DMSO.

  • Available in high purity, typically >99%
  • Used as an anti-thyroid agent
  • Blocks thyroid hormone production
  • Packaged as a 10 mM solution in DMSO
  • Store powder at -20°C for up to 3 years or 4°C for 2 years
  • Store solutions in solvent at -80°C for up to 2 years or -20°C for 1 year

Catalog No. 50-004-05773 Supplier Medchemexpress LLC Supplier No. HYB020810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methimazole, also known as Thiamazole, is an anti-thyroid agent that effectively blocks the production of thyroid hormones from the thyroid gland. This compound is known to potentially cause hepatotoxicity. It is provided as a 10mM solution in 1mL of DMSO.

  • Available in high purity, typically >99%
  • Used as an anti-thyroid agent
  • Blocks thyroid hormone production
  • Packaged as a 10 mM solution in DMSO
  • Store powder at -20°C for up to 3 years or 4°C for 2 years
  • Store solutions in solvent at -80°C for up to 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.