Learn More
Apexbio Technology LLC Methimazole 60-56-0 25g
SDP
Catalog No. 5000095769
Encompass_Preferred
Methimazole is a small-molecule inhibitor targeting intercellular adhesion molecule-1 (ICAM-1) gene transcription It is designed to modulate ICAM-1 expression thereby regulating cell adhesion processes mediated by immunoglobulin superfamily members and integrins such as LFA-1 and Mac-1 Methimazole exerts its biological activity primarily through modulation of signal transducer and activator of transcription 1 (STAT1) In cell-based studies using thyroid cell models methimazole demonstrates inhibitory activity suppressing ICAM-1 mRNA expression induced by interferon-gamma (IFN- ) and H O Based on these pharmacological properties methimazole holds research potential in studying autoimmune thyroid conditions and as an agent to induce hypothyroidism in animal models for thyroid dysfunction research
Catalog No. 50-000-95769 Supplier Apexbio Technology LLC Supplier No. A847225000
Methimazole is a small-molecule inhibitor targeting intercellular adhesion molecule-1 (ICAM-1) gene transcription It is designed to modulate ICAM-1 expression thereby regulating cell adhesion processes mediated by immunoglobulin superfamily members and integrins such as LFA-1 and Mac-1 Methimazole exerts its biological activity primarily through modulation of signal transducer and activator of transcription 1 (STAT1) In cell-based studies using thyroid cell models methimazole demonstrates inhibitory activity suppressing ICAM-1 mRNA expression induced by interferon-gamma (IFN- ) and H O Based on these pharmacological properties methimazole holds research potential in studying autoimmune thyroid conditions and as an agent to induce hypothyroidism in animal models for thyroid dysfunction research