Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methimazole, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111012226 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2226 100 g
11-101-2225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2226 Supplier MilliporeSigma Supelco Supplier No. M8506100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methimazole is an antithyroid agent, widely used in the treatment of hyperthyroidism.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methimazole
CAS 60-56-0
Physical Form Neat
Linear Formula C4H6N2 S
Technique Gas Chromatography
MDL Number MFCD00179321
Melting Point 144°C to 147°C
Synonym 1-Methyl-2-imidazolethiol; 2-Mercapto-1-methylimidazole
Type Analytical Standard
Solvent or Matrix Neat
Industry Type Pharmaceutical
Molecular Weight (g/mol) 114.17
Quantity 100 g
Formula Weight 114.17
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.