Learn More
Cayman Chemical MethImazole-d3 500ug
SDP
Catalog No. 502948738
Encompass_Preferred
An internal standard for the quantification of methimazole by GC- or LC-MS
Catalog No. 50-294-8738 Supplier Cayman Chemical Supplier No. 33278500 UG
An internal standard for the quantification of methimazole by GC- or LC-MS