Learn More
Cayman Chemical MethIocarb-d3 1mg
SDP
Catalog No. 502939313
Encompass_Preferred
An internal standard for the quantification of methiocarb by GC- or LC-MS
Catalog No. 50-293-9313 Supplier Cayman Chemical Supplier No. 277761 MG
Add to Cart
Add to Cart
An internal standard for the quantification of methiocarb by GC- or LC-MS