Learn More
Cayman Chemical Methiocarb-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1581694-94-1, 5 mg
SDP
Catalog No. 502937347
Encompass_Preferred
An internal standard for the quantification of methiocarb by GC- or LC-MS
Catalog No. 50-293-7347 Supplier Cayman Chemical Supplier No. 277765 MG
An internal standard for the quantification of methiocarb by GC- or LC-MS