Learn More
Cayman Chemical MethIonIn SulfoxID IgGBloc 1ea
SDP
Catalog No. 502894177
Encompass_Preferred
A histamine H2 receptor agonist induces acid secretion in isolated human gastric glands (EC50 120 UM) increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg
Catalog No. 50-289-4177 Supplier Cayman Chemical Supplier No. 6001631 EA
A histamine H2 receptor agonist induces acid secretion in isolated human gastric glands (EC50 120 UM) increases pepsin and acid levels in gastric juice in dogs at 3-10 mg/kg