Learn More
Medchemexpress LLC Methionine-3,3,4,4-d4 | 93709-61-6 | 98.0% | 153.24 g/mol | C5H7D4NO2S | 10 MG

Supplier: Medchemexpress LLC HYN0325S410MG
DL-Methionine-d4 is the deuterium-labeled form of the essential amino acid DL-methionine used as a stable-isotope tracer and internal standard in analytical and biochemical workflows. It is supplied as a white to off-white solid with high isotopic enrichment and chemical purity, suitable for LC-MS and isotope-dilution methods.
- Chemical name: Methionine-3,3,4,4-d4.
- Cas number: 93709-61-6.
- Purity: 98.0%.
- Isotopic enrichment: 99.10%.
- Physical form: White to off-white solid.
- Molecular formula: C5H7D4NO2S; molecular weight: 153.24 g/mol.
- Applications: Stable-isotope tracing and internal standard for LC-MS and analytical assays.
- Storage recommendations: Powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.