Learn More
Apexbio Technology LLC Methionine 348-67-4 500mg
SDP
Catalog No. 5000092692
Encompass_Preferred
Methionine (CAS 348-67-4) is a sulfur-containing essential amino acid classified among methionine and its derivatives characterized by modifications at the amino or carboxyl groups or heteroatom substitutions As a precursor in the biosynthesis of S-adenosylmethionine (SAM) methionine donates methyl groups for numerous methylation reactions central to gene expression protein function and metabolic regulation In biomedical research methionine is utilized to investigate mechanisms of transmethylation oxidative stress response and cellular proliferation as well as in studies of amino acid metabolism and epigenetic modulation
Catalog No. 50-000-92692 Supplier Apexbio Technology LLC Supplier No. C6790500
Methionine (CAS 348-67-4) is a sulfur-containing essential amino acid classified among methionine and its derivatives characterized by modifications at the amino or carboxyl groups or heteroatom substitutions As a precursor in the biosynthesis of S-adenosylmethionine (SAM) methionine donates methyl groups for numerous methylation reactions central to gene expression protein function and metabolic regulation In biomedical research methionine is utilized to investigate mechanisms of transmethylation oxidative stress response and cellular proliferation as well as in studies of amino acid metabolism and epigenetic modulation