Learn More
Revvity Health Sciences Inc Methionine, Cell Labeling Grade, L-[35S]-,>600 Ci (22.2 TBq)/mmol, 50 mM Tricine, 10 mM BME, 10 mCi
Catalog No. 502812388
Encompass
Methionine, Cell Labeling Grade, L-[35S]-,>600 Ci (22.2 TBq)/mmol, 50 mM Tricine, 10 mM BME, 10 mCi
Catalog No. 50-281-2388 Supplier Revvity Health Sciences Inc Supplier No. NEG009L010MC
Methionine, Cell Labeling Grade, L-[35S]-,>600 Ci (22.2 TBq)/mmol, 50 mM Tricine, 10 mM BME, 10 mCi