Learn More
Description
- Methionine/choline control diet (MCCD) triggers the resolution of hepatic inflammatory and fibrotic reactions and hepatocyte apoptosis, suggesting that MCDD-induced steatohepatitis is also reversible
- CD diet is used in Non-alcoholic steatohepatitis study
Specifications
Specifications
| Product Type | Animal Diet |
| Format | Pellets |
| Quantity | 10 kg |
| Storage Requirements | Room Temperature |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.