Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Methionine/Choline Control Diet, Pellet

Catalog No. MP296044120 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP296044120 20 kg
MP296044110 10 kg
MP296044150 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP296044120 Supplier MP Biomedicals™ Supplier No. 0296044120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the study of non-alcoholic steatohepatitis disease

  • Methionine/choline control diet (MCCD) triggers the resolution of hepatic inflammatory and fibrotic reactions and hepatocyte apoptosis, suggesting that MCDD-induced steatohepatitis is also reversible
  • CD diet is used in Non-alcoholic steatohepatitis study
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Pellets
Quantity 20 kg
Storage Requirements Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.