Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Methionine/Choline Deficient Diet, Pellet

Catalog No. MP296043910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP296043910 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP296043910 Supplier MP Biomedicals™ Supplier No. 0296043910
Add to Cart
Add to Cart

Methionine/Choline Deficient Diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.

  • Non-alcoholic steatohepatitis(NASH) was induced by feeding a methionine/choline-deficient diet (MCD-diet) for up to 5 weeks to mice
  • NASH is a disease with a high incidence, difficult diagnosis, and as yet no effective treatment
  • So,MCD-diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Pellets
Quantity 10 kg
Storage Requirements Store at Room Temperature(15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.