Learn More
Description
- Contains sucrose, corn starch, corn oil, Alphacel non-nutritive bulk, and AIN 76 mineral mix.
Specifications
Specifications
| Format | Powder |
| Quantity | 10 kg |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.