Learn More
Medchemexpress LLC Methionine-d4 | 00-00-0 | 98.0% | 153.24 g/mol | C5H7D4NO2S | 5 MG
SDP
Catalog No. 5000221958
Encompass_Preferred
Methionine-d4 is a deuterium-labeled analogue of methionine used as an internal standard and tracer in analytical and biochemical workflows. It is intended for quantitative mass spectrometry, metabolic tracing, and experiments that require isotopically labeled amino acids. Supplied in small pack sizes with high stated purity.
Catalog No. 50-002-21958 Supplier Medchemexpress LLC Supplier No. HY13694S15MG
Methionine-d4 is a deuterium-labeled analogue of methionine used as an internal standard and tracer in analytical and biochemical workflows. It is intended for quantitative mass spectrometry, metabolic tracing, and experiments that require isotopically labeled amino acids. Supplied in small pack sizes with high stated purity.