Learn More
Revvity Health Sciences Inc Methionine, L-[35S]-, 1mCi (37MBq), Specific Activity:>1000Ci (37.0TBq)/mmol, 10mM BME
Catalog No. 502809820
Encompass
Methionine, L-[35S]-, 1mCi (37MBq), Specific Activity:>1000Ci (37.0TBq)/mmol, 10mM BME
Catalog No. 50-280-9820 Supplier Revvity Health Sciences Inc Supplier No. NEG009T001MC
Add to Cart
Add to Cart
Methionine, L-[35S]-, 1mCi (37MBq), Specific Activity:>1000Ci (37.0TBq)/mmol, 10mM BME