Learn More
Medchemexpress LLC HY-W010104A 500mg Medchemexpress, Methionine sulfoxide CAS:62697-73-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010104A500MG
Encompass_Preferred
Medchemexpress, HY-W010104A 500mg Methionine sulfoxide CAS:62697-73-8 Methionine sulfoxide is an oxidation product of methionine with reactive oxygen species and can be regarded as a biomarker of oxidative stress in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3282