Learn More
Medchemexpress LLC Methionine sulfoximine | 1982-67-8 | MFCD00063094 | 99.6% | 180.23 | C5H12N2O3S | 25 MG
SDP
Catalog No. 5000282350
Encompass_Preferred
Methionine sulfoximine is an irreversible glutamine synthetase inhibitor used as a research tool to probe glutamate and glutamine metabolism and to study seizure-related mechanisms. Supplied as a 25 mg sample with high analytical purity, it is intended for research and laboratory applications.
Catalog No. 50-002-82350 Supplier Medchemexpress LLC Supplier No. HYW33814025MG
Methionine sulfoximine is an irreversible glutamine synthetase inhibitor used as a research tool to probe glutamate and glutamine metabolism and to study seizure-related mechanisms. Supplied as a 25 mg sample with high analytical purity, it is intended for research and laboratory applications.