Learn More
Medchemexpress LLC Methionine sulfoximine (2-amino-4-(S-methylsulfonimidoyl)butanoic acid) | 1982-67-8 | MFCD00002621 | 99.6% | 180.23 g/mol | C5H12N2O3S | 5 MG
SDP
Catalog No. 5000282558
Encompass_Preferred
Methionine sulfoximine is a small-molecule biochemical reagent and an irreversible inhibitor of glutamine synthetase. It modulates glutamate and glutamine metabolism and is used in neurobiological research, including studies of seizure mechanisms and glutamine-dependent pathways. Supplied as a high-purity solid suitable for laboratory research applications.
  • Irreversible inhibitor of glutamine synthetase.
  • Modulates glutamate and glutamine metabolism.
  • Used in neurobiology and seizure-related research.
  • Supplied as a high-purity reagent suitable for analytical and experimental use.
  • Available in small milligram- and gram-scale pack sizes for laboratory workflows.
Catalog No. 50-002-82558 Supplier Medchemexpress LLC Supplier No. HYW3381405MG
Methionine sulfoximine is a small-molecule biochemical reagent and an irreversible inhibitor of glutamine synthetase. It modulates glutamate and glutamine metabolism and is used in neurobiological research, including studies of seizure mechanisms and glutamine-dependent pathways. Supplied as a high-purity solid suitable for laboratory research applications.
  • Irreversible inhibitor of glutamine synthetase.
  • Modulates glutamate and glutamine metabolism.
  • Used in neurobiology and seizure-related research.
  • Supplied as a high-purity reagent suitable for analytical and experimental use.
  • Available in small milligram- and gram-scale pack sizes for laboratory workflows.