Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methionine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604388 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-388 25 kg Amber Glass Bottle
18-604-384 25 g Amber Glass Bottle
18-604-385 100 g Amber Glass Bottle
18-604-386 500 g Amber Glass Bottle
18-604-387 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-388 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M144125KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1441, 63-68-3, C5H11NO2S

Spectrum™ Chemical Methionine, USP is used to prevent liver damage in acetaminophen poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.4%
Loss on Drying 0.3%
Packaging Amber Glass Bottle
Quantity 25 kg
Specific Rotation +22.4° to +24.7°
Percent Purity 98.5 to 101.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.