Learn More
Creative Biomart Somatrem; Recombinant HGH
Catalog No. 502084086
Encompass_Preferred
The protein has the same amino acid sequence as human growth hormone except for an extra methionine at the end of the sequence chain to facilitate its manufacturing process.
Catalog No. 50-208-4086 Supplier Creative Biomart Supplier No. THP0035
The protein has the same amino acid sequence as human growth hormone except for an extra methionine at the end of the sequence chain to facilitate its manufacturing process.