Learn More
Apexbio Technology LLC Methiothepin maleate 19728-88-2 50mg
SDP
Catalog No. 5000092059
Encompass_Preferred
Methiothepin maleate (CAS 19728-88-2) is a small molecule antagonist targeting serotonin receptors including 5-HT1 5-HT6 and 5-HT7 subtypes By inhibiting serotonin autoreceptors it modulates downstream signaling pathways associated with neurotransmission This compound has been widely utilized in neuroscience research to investigate the regulatory mechanisms of serotonergic signaling and to explore the pathophysiology of related neurological disorders Its ability to selectively block multiple serotonin receptor subtypes makes it a valuable tool for delineating complex neural pathways and receptor functions in experimental models
Catalog No. 50-000-92059 Supplier Apexbio Technology LLC Supplier No. B636250
Methiothepin maleate (CAS 19728-88-2) is a small molecule antagonist targeting serotonin receptors including 5-HT1 5-HT6 and 5-HT7 subtypes By inhibiting serotonin autoreceptors it modulates downstream signaling pathways associated with neurotransmission This compound has been widely utilized in neuroscience research to investigate the regulatory mechanisms of serotonergic signaling and to explore the pathophysiology of related neurological disorders Its ability to selectively block multiple serotonin receptor subtypes makes it a valuable tool for delineating complex neural pathways and receptor functions in experimental models