Learn More
Cayman Chemical MethIothepInmaleate 100mg
SDP
Catalog No. 502995082
Encompass_Preferred
A DP2/CRTH2 receptor antagonist (IC50 3 98 nM) selective for DP2 over a panel of receptors ion channels transporters and enzymes including DP1 COX-1 and COX-2 at 10 UM inhibits 1314-dihydro-15-keto-PGD2-induced increases in CD11b expression in isolated human eosinophils inhibits 1314-dihydro-15-keto-PGD2-induced chemotaxis of isolated human eosinophils and T cells in a concentration-dependent manner
Catalog No. 50-299-5082 Supplier Cayman Chemical Supplier No. 23138100 MG
A DP2/CRTH2 receptor antagonist (IC50 3 98 nM) selective for DP2 over a panel of receptors ion channels transporters and enzymes including DP1 COX-1 and COX-2 at 10 UM inhibits 1314-dihydro-15-keto-PGD2-induced increases in CD11b expression in isolated human eosinophils inhibits 1314-dihydro-15-keto-PGD2-induced chemotaxis of isolated human eosinophils and T cells in a concentration-dependent manner