Learn More
Cayman Chemical MethIothepInmaleate 10mg
SDP
Catalog No. 502995080
Encompass_Preferred
A fluorescent membrane-permeable biarsenical compound that binds covalently to tetracysteine sequences which are engineered into target proteins
Catalog No. 50-299-5080 Supplier Cayman Chemical Supplier No. 2313810 MG
A fluorescent membrane-permeable biarsenical compound that binds covalently to tetracysteine sequences which are engineered into target proteins