Learn More
STEMCELL Technologies MethoCult™ GF M3434, Size: 100 mL
SDP
Catalog No. NC9772657
Encompass
MethoCult™ GF M3434 is optimized for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells. MethoCult™ GF M3434 has been formulated to support optimal growth of primitive erythroid progenitor cells (BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM). This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.
Catalog No. NC9772657 Supplier STEMCELL Technologies Supplier No. 03434
MethoCult™ GF M3434 is optimized for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of mouse bone marrow, spleen, peripheral blood, and fetal liver cells. MethoCult™ GF M3434 has been formulated to support optimal growth of primitive erythroid progenitor cells (BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM). This formulation is compatible with STEMvision™ software for automated colony counting of mouse bone marrow CFU assays.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/technical-resources/educational-materials/tech-tips/frequently-asked-questions-cfu-assay.html) on performing the CFU assay.