Learn More
Medchemexpress LLC HY-B0262 500mg Medchemexpress, Methocarbamol CAS:532-03-6 Purity:>98%
SDP
Catalog No. 502082432
Encompass_Preferred
Medchemexpress, HY-B0262 500mg Methocarbamol CAS:532-03-6 Methocarbamol is an orally active central muscle relaxant and blocks muscular Nav1.4 channel. Methocarbamol reversibly affects voltage dependence of inactivation of Nav1.4 channel. Methocarbamol has the potential for muscle spasms and pain syndromes research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2432 Supplier Medchemexpress LLC Supplier No. HYB0262500MG
Medchemexpress, HY-B0262 500mg Methocarbamol CAS:532-03-6 Methocarbamol is an orally active central muscle relaxant and blocks muscular Nav1.4 channel. Methocarbamol reversibly affects voltage dependence of inactivation of Nav1.4 channel. Methocarbamol has the potential for muscle spasms and pain syndromes research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.