Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methocarbamol | 532-03-6 | 99.8% | 10 MM 1 ML
SDP

Catalog No. 5000405659
Encompass_Preferred

Methocarbamol is an orally active central muscle relaxant that blocks the muscular Nav1.4 channel. It reversibly affects the voltage dependence of inactivation of the Nav1.4 channel. This compound has potential for research into muscle spasms and pain syndromes. It is intended for research use only and not for patient administration.

  • Blocks muscular Nav1.4 channel
  • Reversibly affects voltage dependence of inactivation of Nav1.4 channel
  • Potential for research into muscle spasms and pain syndromes
  • Orally active central muscle relaxant
  • Purity of 99.8%
  • Molecular formula: C11H15NO5
  • Molecular weight: 241.2405
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-05659 Supplier Medchemexpress LLC Supplier No. HYB026210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methocarbamol is an orally active central muscle relaxant that blocks the muscular Nav1.4 channel. It reversibly affects the voltage dependence of inactivation of the Nav1.4 channel. This compound has potential for research into muscle spasms and pain syndromes. It is intended for research use only and not for patient administration.

  • Blocks muscular Nav1.4 channel
  • Reversibly affects voltage dependence of inactivation of Nav1.4 channel
  • Potential for research into muscle spasms and pain syndromes
  • Orally active central muscle relaxant
  • Purity of 99.8%
  • Molecular formula: C11H15NO5
  • Molecular weight: 241.2405
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.