Learn More
Medchemexpress LLC Methocarbamol | 532-03-6 | MFCD00057662 | 99.8% | 100 MG
SDP
Catalog No. 5000406954
Encompass_Preferred
Methocarbamol is an orally active central muscle relaxant that blocks the muscular Nav1.4 channel. It reversibly affects voltage dependence of inactivation of the Nav1.4 channel, making it suitable for research into muscle spasms and pain syndromes.
  • Acts as an orally active central muscle relaxant
  • Blocks muscular Nav1.4 channels
  • Reversibly affects voltage dependence of inactivation of Nav1.4 channel
  • Significantly increases decay times of EPCs and EPPs induced by phrenic nerve stimulation
  • Demonstrates muscle relaxant activity in vivo
Catalog No. 50-004-06954 Supplier Medchemexpress LLC Supplier No. HYB0262100MG
Methocarbamol is an orally active central muscle relaxant that blocks the muscular Nav1.4 channel. It reversibly affects voltage dependence of inactivation of the Nav1.4 channel, making it suitable for research into muscle spasms and pain syndromes.
  • Acts as an orally active central muscle relaxant
  • Blocks muscular Nav1.4 channels
  • Reversibly affects voltage dependence of inactivation of Nav1.4 channel
  • Significantly increases decay times of EPCs and EPPs induced by phrenic nerve stimulation
  • Demonstrates muscle relaxant activity in vivo