Learn More
Cayman Chemical Methocarbamol, Purity: ≥98%, CAS: 532-03-6, 5 g
SDP
Catalog No. 502994835
Encompass_Preferred
An orally bioavailable skeletal muscle relaxant; inhibits the ability of mice to remain on a vertical ladder for 1 minute (ED50 = 15 mg/kg) and decreases forelimb grip strength by 35.9% (500 mg/kg); abolishes femoral nerve-stimulated polysynaptic reflex contractions of the cat tibialis anterior muscle and prolongs the mean refractory period of directly or indirectly stimulated skeletal muscle (200 mg/kg); inhibits human CAI over CAII (IC50s = 70 and ~80,000 μM, respectively)
Catalog No. 50-299-4835 Supplier Cayman Chemical Supplier No. 238705 G
An orally bioavailable skeletal muscle relaxant; inhibits the ability of mice to remain on a vertical ladder for 1 minute (ED50 = 15 mg/kg) and decreases forelimb grip strength by 35.9% (500 mg/kg); abolishes femoral nerve-stimulated polysynaptic reflex contractions of the cat tibialis anterior muscle and prolongs the mean refractory period of directly or indirectly stimulated skeletal muscle (200 mg/kg); inhibits human CAI over CAII (IC50s = 70 and ~80,000 μM, respectively)