Learn More
Medchemexpress LLC Methocarbamol-d3 | 1346600-86-9 | 98.2% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0262S11MG
Encompass_Preferred
Methocarbamol-d3 is a deuterium-labeled form of Methocarbamol, an orally active central muscle relaxant. It functions by blocking the muscular Nav1.4 channel and reversibly influences the voltage dependence of its inactivation. It is valuable for research into muscle spasms and pain syndromes.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06911